Search
Search

afp4| Gene Symbol: afp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045488.1 | NP_001038953.1 | 2 - 3 | 100 | 69 | |
| GenBank mRNA | AY345904.1 | - | 2 - 3 | 100 | 69 | |
| BC133822.1 | - | 1 - 2 | 64 | 69 | ||
| KJ183061.1 | - | 2 - 3 | 101 | 69 | ||
| KM655710.1 | - | 1 - 2 | 46 | 69 | ||
Set Membership: |
|