Search
Search

phkg1a| Gene Symbol: phkg1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077612.1 | NP_001071080.1 | 9 - 10 | 1084 | 62 | |
| GenBank mRNA | BC125847.1 | - | 9 - 10 | 1084 | 62 | |
| BC164489.1 | - | 8 - 9 | 952 | 62 | ||
Set Membership: |
|