Search
Search

satb1a| Gene Symbol: satb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080162.1 | NP_001073631.1 | 4 - 5 | 729 | 80 | |
| XM_005170006.4 | XP_005170063.1 | 4 - 5 | 824 | 80 | ||
| GenBank mRNA | BC163787.1 | - | 4 - 5 | 748 | 80 | |
| BC163793.1 | - | 4 - 5 | 748 | 80 | ||
Set Membership: |
|