Search
Search

cfap45| Gene Symbol: cfap45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082996.2 | NP_001076465.2 | 11 - 12 | 1748 | 77 | |
| GenBank mRNA | BC133121.1 | - | 11 - 12 | 1748 | 77 | |
Set Membership: |
|