Search
Search

leng8| Gene Symbol: leng8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089519.1 | NP_001082988.1 | 1 - 2 | 250 | 70 | |
| XM_009292081.3 | XP_009290356.1 | 2 - 3 | 188 | 70 | ||
| GenBank mRNA | BC139488.1 | - | 1 - 2 | 250 | 70 | |
Set Membership: |
|