Search
Search

spint1b| Gene Symbol: spint1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111223.1 | NP_001104693.1 | 9 - 10 | 1529 | 104 | |
| GenBank mRNA | BC155551.1 | - | 9 - 10 | 1529 | 104 | |
Set Membership: |
|