Search
Search

pi4kab| Gene Symbol: pi4kab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113342.1 | NP_001106813.1 | 14 - 15 | 1725 | 71 | |
| GenBank mRNA | BC155592.1 | - | 14 - 15 | 1725 | 71 | |
Set Membership: |
|