Search
Search

ipp| Gene Symbol: ipp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113621.1 | NP_001107093.1 | 8 - 9 | 1492 | 121 | |
| XM_017358910.2 | XP_017214399.1 | 8 - 9 | 2265 | 121 | ||
| XM_017358916.2 | XP_017214405.1 | 6 - 7 | 856 | 121 | ||
| XM_017358919.2 | XP_017214408.1 | 8 - 9 | 2284 | 121 | ||
| GenBank mRNA | BC154444.1 | - | 8 - 9 | 1492 | 121 | |
| BC171521.1 | - | 8 - 9 | 1487 | 121 | ||
Set Membership: |
|