Search
Search

heatr6| Gene Symbol: heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114313.1 | NP_001107785.1 | 9 - 10 | 1451 | 62 | |
| GenBank mRNA | BC155667.1 | - | 9 - 10 | 1451 | 62 | |
Set Membership: |
|