Search
Search

myb| Gene Symbol: myb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309822.1 | NP_001296751.1 | 4 - 5 | 471 | 71 | |
| NM_131266.2 | NP_571341.2 | 4 - 5 | 471 | 71 | ||
| GenBank mRNA | AF191559.1 | - | 4 - 5 | 402 | 71 | |
| BC059803.1 | - | 4 - 5 | 326 | 71 | ||
Set Membership: |
|