Search
Search

coil| Gene Symbol: coil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131512.2 | NP_571587.2 | 2 - 3 | 1360 | 134 | |
| GenBank mRNA | AF220008.1 | - | 1 - 2 | 158 | 134 | |
| BC045858.1 | - | 2 - 3 | 1261 | 134 | ||
| BC164689.1 | - | 2 - 3 | 1266 | 134 | ||
| EH566753.1 | - | 5 - 6 | 746 | 134 | ||
Set Membership: |
|