Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: tbx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153665.1 | NP_705951.1 | 4 - 5 | 891 | 103 | |
GenBank mRNA | AF448503.3 | - | 4 - 5 | 879 | 103 | |
BC154826.1 | - | 4 - 5 | 875 | 103 | ||
BC163460.1 | - | 4 - 5 | 836 | 103 | ||
BC163482.1 | - | 4 - 5 | 836 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |