Search
Search

glula| Gene Symbol: glula | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181559.2 | NP_853537.1 | 2 - 3 | 276 | 85 | |
| GenBank mRNA | BC045255.1 | - | 2 - 3 | 274 | 85 | |
| BC053146.1 | - | 2 - 3 | 261 | 85 | ||
| BC171365.1 | - | 1 - 2 | 192 | 85 | ||
| BC171369.1 | - | 2 - 3 | 197 | 85 | ||
| EH438060.1 | - | 2 - 3 | 290 | 85 | ||
Set Membership: |
|