Search
Search

brf1a| Gene Symbol: brf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199898.1 | NP_956192.1 | 9 - 10 | 1109 | 122 | |
| XM_017358699.2 | XP_017214188.1 | 9 - 10 | 1165 | 122 | ||
| GenBank mRNA | BC046877.1 | - | 9 - 10 | 1109 | 122 | |
| BC165640.1 | - | 8 - 9 | 950 | 122 | ||
Set Membership: |
|