Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: desi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200208.1 | NP_956502.1 | 4 - 5 | 561 | 123 | |
GenBank mRNA | BC045987.1 | - | 4 - 5 | 561 | 123 | |
BC171711.1 | - | 4 - 5 | 512 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |