Search
Search

siae| Gene Symbol: siae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044331.1 | NP_001037796.1 | 9 - 10 | 1321 | 76 | |
| GenBank mRNA | BC049448.1 | - | 9 - 10 | 1321 | 76 | |
| BC134862.1 | - | 9 - 10 | 1293 | 76 | ||
Set Membership: |
|