Search
Search

prom1a| Gene Symbol: prom1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115143.2 | NP_001108615.2 | 19 - 20 | 2294 | 125 | |
| XM_005157347.3 | XP_005157404.1 | 19 - 20 | 2419 | 125 | ||
| XM_005157349.3 | XP_005157406.1 | 21 - 22 | 2471 | 125 | ||
| XM_005157351.3 | XP_005157408.1 | 20 - 21 | 2447 | 125 | ||
| XM_009291267.3 | XP_009289542.1 | 21 - 22 | 2573 | 125 | ||
| XM_009291269.3 | XP_009289544.1 | 19 - 20 | 2419 | 125 | ||
| XM_021480968.1 | XP_021336643.1 | 22 - 23 | 2592 | 125 | ||
| XM_021480969.1 | XP_021336644.1 | 21 - 22 | 2568 | 125 | ||
| XM_021480970.1 | XP_021336645.1 | 21 - 22 | 2565 | 125 | ||
| XM_021480971.1 | XP_021336646.1 | 22 - 23 | 2591 | 125 | ||
| XM_021480972.1 | XP_021336647.1 | 22 - 23 | 2591 | 125 | ||
| XM_021480973.1 | XP_021336648.1 | 20 - 21 | 2438 | 125 | ||
| XM_021480974.1 | XP_021336649.1 | 22 - 23 | 2592 | 125 | ||
| XM_021480975.1 | XP_021336650.1 | 22 - 23 | 2591 | 125 | ||
| GenBank mRNA | BC163446.1 | - | 19 - 20 | 2294 | 125 | |
Set Membership: |
|