Search
Search

amd1| Gene Symbol: amd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161601.1 | NP_001155073.1 | 2 - 3 | 441 | 91 | |
| GenBank mRNA | BC045404.1 | - | 2 - 3 | 420 | 91 | |
| BC163959.1 | - | 2 - 3 | 238 | 91 | ||
Set Membership: |
|