Search
Search

pmp22a| Gene Symbol: pmp22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201311.1 | NP_958468.1 | 3 - 4 | 408 | 117 | |
| XM_005164439.4 | XP_005164496.1 | 2 - 3 | 158 | 117 | ||
| XM_005164440.4 | XP_005164497.1 | 2 - 3 | 249 | 117 | ||
| GenBank mRNA | BC062526.1 | - | 3 - 4 | 408 | 117 | |
| BC153614.1 | - | 2 - 3 | 212 | 117 | ||
Set Membership: |
|