Search
Search

ikzf5| Gene Symbol: ikzf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205601.1 | NP_991164.1 | 2 - 3 | 509 | 85 | |
| XM_009293021.3 | XP_009291296.1 | 2 - 3 | 532 | 85 | ||
| GenBank mRNA | AY394931.1 | - | 2 - 3 | 509 | 85 | |
| BC078341.1 | - | 1 - 2 | 183 | 85 | ||
Set Membership: |
|