Search
Search

ttn.1| Gene Symbol: ttn.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021479071.1 | XP_021334746.1 | 209 - 210 | 83107 | 149 | |
| XM_021479072.1 | XP_021334747.1 | 167 - 168 | 75506 | 149 | ||
| GenBank mRNA | BC095863.1 | - | 2 - 3 | 439 | 149 | |
| BC115205.1 | - | 2 - 3 | 466 | 149 | ||
| BC125897.1 | - | 2 - 3 | 511 | 149 | ||
| CK677667.1 | - | 1 - 2 | 169 | 149 | ||
| CK694327.1 | - | 2 - 3 | 443 | 149 | ||
Set Membership: |
|