Search
Search

slc16a3| Gene Symbol: slc16a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212708.1 | NP_997873.1 | 2 - 3 | 277 | 105 | |
| XM_005169469.4 | XP_005169526.1 | 2 - 3 | 472 | 105 | ||
| GenBank mRNA | AB610608.1 | - | 1 - 2 | 251 | 105 | |
| BC045489.1 | - | 2 - 3 | 277 | 105 | ||
| BC154231.1 | - | 2 - 3 | 277 | 105 | ||
Set Membership: |
|