Search
Search

srsf9| Gene Symbol: srsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312899.1 | NP_001299828.1 | 4 - 5 | 681 | 69 | |
| NM_212899.2 | NP_998064.2 | 4 - 5 | 859 | 69 | ||
| GenBank mRNA | BC067134.1 | - | 4 - 5 | 785 | 69 | |
| BC154583.1 | - | 4 - 5 | 600 | 69 | ||
| EH464318.1 | - | 1 - 2 | 636 | 69 | ||
| EH504229.1 | - | 4 - 5 | 662 | 69 | ||
| EH583693.1 | - | 4 - 5 | 803 | 69 | ||
Set Membership: |
|