Search
Search

hnrpkl| Gene Symbol: hnrpkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212991.3 | NP_998156.3 | 10 - 11 | 1048 | 117 | |
| GenBank mRNA | BC096867.1 | - | 10 - 11 | 1028 | 117 | |
| BC164732.1 | - | 9 - 10 | 970 | 117 | ||
Set Membership: |
|