Search
Search

vcanb| Gene Symbol: vcanb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214688.1 | NP_999853.1 | 10 - 11 | 4059 | 68 | |
| GenBank mRNA | AB119257.1 | - | 10 - 11 | 4059 | 68 | |
| BC162605.1 | - | 10 - 11 | 4045 | 68 | ||
Set Membership: |
|