Search
Search

bsx| Gene Symbol: bsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214727.1 | NP_999892.1 | 2 - 3 | 526 | 81 | |
| GenBank mRNA | AY515251.1 | - | 2 - 3 | 526 | 81 | |
| BC162183.1 | - | 2 - 3 | 501 | 81 | ||
| BC162209.1 | - | 2 - 3 | 501 | 81 | ||
Set Membership: |
|