Search
Search

WDFY2| Gene Symbol: WDFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732071.2 | XP_014587557.1 | 1066 | 77 | ||
| XM_023621542.1 | XP_023477310.1 | 1720 | 77 | |||
| XM_023621543.1 | XP_023477311.1 | 4098 | 77 | |||
| XM_023621544.1 | XP_023477312.1 | 747 | 77 | |||
| GenBank mRNA | JL628146.1 | - | 4 - 5 | 860 | 77 | |
Set Membership: |
|