Search
Search
| Gene Symbol: OGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493372.5 | XP_001493422.1 | 687 | 80 | ||
| XM_001493388.5 | XP_001493438.1 | 657 | 80 | |||
| XM_005614262.3 | XP_005614319.1 | 534 | 80 | |||
Set Membership: |
|