Search
Search

PDS5A| Gene Symbol: PDS5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005608813.3 | XP_005608870.1 | 2639 | 61 | ||
| XM_023638209.1 | XP_023493977.1 | 2817 | 61 | |||
| XM_023638210.1 | XP_023493978.1 | 2700 | 61 | |||
| XM_023638211.1 | XP_023493979.1 | 2639 | 61 | |||
| XM_023638212.1 | XP_023493980.1 | 2628 | 61 | |||
| XM_023638213.1 | XP_023493981.1 | 2639 | 61 | |||
Set Membership: |
|