Search
Search

ZNF462| Gene Symbol: ZNF462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493275.6 | XP_001493325.1 | 6136 | 69 | ||
| XM_014735941.2 | XP_014591427.1 | 6136 | 69 | |||
| XM_014735943.2 | XP_014591429.1 | 6136 | 69 | |||
| XM_023629835.1 | XP_023485603.1 | 7170 | 69 | |||
| XM_023629836.1 | XP_023485604.1 | 6634 | 69 | |||
| XM_023629837.1 | XP_023485605.1 | 6258 | 69 | |||
| XM_023629838.1 | XP_023485606.1 | 7873 | 69 | |||
| XM_023629839.1 | XP_023485607.1 | 6139 | 69 | |||
| XM_023629840.1 | XP_023485608.1 | 6141 | 69 | |||
Set Membership: |
|