Search
Search
| Gene Symbol: FGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163886.1 | NP_001157358.1 | 1 - 2 | 209 | 104 | |
| XM_014730308.2 | XP_014585794.1 | 863 | 104 | |||
| XM_023616891.1 | XP_023472659.1 | 626 | 104 | |||
| XM_023616892.1 | XP_023472660.1 | 1033 | 104 | |||
| XM_023616893.1 | XP_023472661.1 | 877 | 104 | |||
| XM_023616894.1 | XP_023472662.1 | 2696 | 104 | |||
| XM_023616895.1 | XP_023472663.1 | 616 | 104 | |||
| XM_023616896.1 | XP_023472664.1 | 623 | 104 | |||
| GenBank mRNA | AJ439890.1 | - | 1 - 2 | 119 | 104 | |
Set Membership: |
|