Search
Search

TIAL1| Gene Symbol: TIAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638358.1 | XP_023494126.1 | 431 | 77 | ||
| XM_023638364.1 | XP_023494132.1 | 380 | 77 | |||
| XM_023638369.1 | XP_023494137.1 | 2010 | 77 | |||
| XM_023638378.1 | XP_023494146.1 | 1957 | 77 | |||
| XM_023638385.1 | XP_023494153.1 | 2039 | 77 | |||
| XM_023638390.1 | XP_023494158.1 | 431 | 77 | |||
| XM_023638395.1 | XP_023494163.1 | 175 | 77 | |||
Set Membership: |
|