Search
Search

PLA2G6| Gene Symbol: PLA2G6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736830.2 | XP_014592316.2 | 2493 | 54 | ||
| XM_023631596.1 | XP_023487364.1 | 2583 | 54 | |||
| XM_023631598.1 | XP_023487366.1 | 2538 | 54 | |||
| XM_023631599.1 | XP_023487367.1 | 3058 | 54 | |||
| XM_023631600.1 | XP_023487368.1 | 2450 | 54 | |||
| XM_023631601.1 | XP_023487369.1 | 2455 | 54 | |||
| XM_023631602.1 | XP_023487370.1 | 2351 | 54 | |||
| XM_023631603.1 | XP_023487371.1 | 2925 | 54 | |||
| XM_023631604.1 | XP_023487372.1 | 2331 | 54 | |||
Set Membership: |
|