Search
Search
| Gene Symbol: BRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496232.4 | XP_001496282.1 | 2871 | 71 | ||
| XM_005603797.3 | XP_005603854.1 | 975 | 71 | |||
| XM_014734338.2 | XP_014589824.1 | 2615 | 71 | |||
| XM_014734339.2 | XP_014589825.1 | 1321 | 71 | |||
| XM_014734340.2 | XP_014589826.1 | 1321 | 71 | |||
| XM_023624692.1 | XP_023480460.1 | 2505 | 71 | |||
| GenBank mRNA | JL627520.1 | - | 1 - 2 | 530 | 71 | |
Set Membership: |
|