Search
Search

UHMK1| Gene Symbol: UHMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005609857.3 | XP_005609914.1 | 1803 | 96 | ||
| XM_005609858.3 | XP_005609915.1 | 1803 | 96 | |||
| XM_005609859.3 | XP_005609916.1 | 1803 | 96 | |||
| XM_023640816.1 | XP_023496584.1 | 674 | 96 | |||
| XM_023640817.1 | XP_023496585.1 | 659 | 96 | |||