Search
Search
| Gene Symbol: ERI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497177.6 | XP_001497227.1 | 441 | 74 | ||
| XM_005607113.3 | XP_005607170.1 | 441 | 74 | |||
| XM_005607114.3 | XP_005607171.1 | 441 | 74 | |||
| XM_023631440.1 | XP_023487208.1 | 441 | 74 | |||
| XM_023631441.1 | XP_023487209.1 | 441 | 74 | |||
Set Membership: |
|