Search
Search
| Gene Symbol: CSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081860.2 | NP_001075329.1 | 3 - 4 | 314 | 97 | |
| NM_001309387.1 | NP_001296316.1 | 4 - 5 | 469 | 97 | ||
| GenBank mRNA | AF503365.1 | - | 3 - 4 | 330 | 97 | |
| BI961242.1 | - | 4 - 5 | 476 | 97 | ||
Set Membership: |
|