Search
Search
| Gene Symbol: ARG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499892.6 | XP_001499942.4 | 947 | 95 | ||
| XM_014735659.2 | XP_014591145.1 | 947 | 95 | |||
| XM_023627973.1 | XP_023483741.1 | 986 | 95 | |||
| XM_023627974.1 | XP_023483742.1 | 704 | 95 | |||
| GenBank mRNA | AY647160.1 | - | 2 - 3 | 186 | 95 | |
| JL640459.1 | - | 7 - 8 | 889 | 95 | ||
Set Membership: |
|