Search
Search

CALB1| Gene Symbol: CALB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163952.1 | NP_001157424.1 | 5 - 6 | 505 | 72 | |
| XM_005613185.3 | XP_005613242.1 | 559 | 72 | |||
| GenBank mRNA | AY665136.1 | - | 3 - 4 | 199 | 72 | |
Set Membership: |
|