Search
Search
| Gene Symbol: SDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734954.2 | XP_014590440.2 | 1640 | 56 | ||
| XM_023625909.1 | XP_023481677.1 | 1637 | 56 | |||
| GenBank mRNA | DQ402987.1 | - | 9 - 10 | 1373 | 56 | |
Set Membership: |
|