Search
Search

KDM5C| Gene Symbol: KDM5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014728918.2 | XP_014584404.1 | 1278 | 64 | ||
| XM_014728919.2 | XP_014584405.1 | 1276 | 64 | |||
| XM_014728920.2 | XP_014584406.1 | 1278 | 64 | |||
| XM_014728921.2 | XP_014584407.1 | 1277 | 64 | |||
| XM_014728922.2 | XP_014584408.1 | 1275 | 64 | |||
| XM_014728923.2 | XP_014584409.1 | 1276 | 64 | |||
| XM_014728924.2 | XP_014584410.1 | 1109 | 64 | |||
| XM_014728925.2 | XP_014584411.1 | 1278 | 64 | |||
| XM_014728926.2 | XP_014584412.1 | 1281 | 64 | |||
| XM_014728927.2 | XP_014584413.1 | 1281 | 64 | |||
| XM_023633551.1 | XP_023489319.1 | 1275 | 64 | |||
| XM_023633552.1 | XP_023489320.1 | 1106 | 64 | |||
| XM_023633553.1 | XP_023489321.1 | 1279 | 64 | |||
| XM_023633554.1 | XP_023489322.1 | 1280 | 64 | |||
| XM_023633555.1 | XP_023489323.1 | 1280 | 64 | |||
| GenBank mRNA | U52363.1 | - | 7 - 8 | 850 | 64 | |
Set Membership: |
|