Search
Search

ADORA2A| Gene Symbol: ADORA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081897.1 | NP_001075366.1 | 2 | 1956 | 95 | |
| XM_023646579.1 | XP_023502347.1 | 2083 | 95 | |||
| GenBank mRNA | AY394857.1 | - | 2 | 1956 | 95 | |
Set Membership: |
|