Search
Search

SPPL3| Gene Symbol: SPPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488416.6 | XP_001488466.4 | 565 | 68 | ||
| XM_023647018.1 | XP_023502786.1 | 586 | 68 | |||
| XM_023647019.1 | XP_023502787.1 | 586 | 68 | |||
| XM_023647020.1 | XP_023502788.1 | 657 | 68 | |||
Set Membership: |
|