Search
Search

PBRM1| Gene Symbol: PBRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600568.3 | XP_005600625.1 | 988 | 76 | ||
| XM_005600569.3 | XP_005600626.1 | 979 | 76 | |||
| XM_005600570.3 | XP_005600627.1 | 968 | 76 | |||
| XM_005600572.3 | XP_005600629.1 | 874 | 76 | |||
| XM_005600573.3 | XP_005600630.1 | 861 | 76 | |||
| XM_023620295.1 | XP_023476063.1 | 773 | 76 | |||
| XM_023620296.1 | XP_023476064.1 | 985 | 76 | |||
| XM_023620297.1 | XP_023476065.1 | 773 | 76 | |||
| XM_023620298.1 | XP_023476066.1 | 893 | 76 | |||
| XM_023620299.1 | XP_023476067.1 | 773 | 76 | |||
| XM_023620300.1 | XP_023476068.1 | 773 | 76 | |||
| XM_023620301.1 | XP_023476069.1 | 773 | 76 | |||
| XM_023620302.1 | XP_023476070.1 | 773 | 76 | |||
| XM_023620303.1 | XP_023476071.1 | 764 | 76 | |||
| XM_023620304.1 | XP_023476072.1 | 773 | 76 | |||
Set Membership: |
|