Search
Search

RBFOX1| Gene Symbol: RBFOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502564.5 | XP_001502614.1 | 3634 | 76 | ||
| XM_023616846.1 | XP_023472614.1 | 1335 | 76 | |||
| XM_023616850.1 | XP_023472618.1 | 1257 | 76 | |||
Set Membership: |
|