Search
Search
| Gene Symbol: CD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252300.1 | NP_001239229.1 | 3 - 4 | 597 | 83 | |
| XM_005609941.3 | XP_005609998.1 | 1115 | 83 | |||
| XM_023640265.1 | XP_023496033.1 | 702 | 83 | |||
| XM_023640266.1 | XP_023496034.1 | 702 | 83 | |||
| GenBank mRNA | JF749875.1 | - | 3 - 4 | 597 | 83 | |
Set Membership: |
|