Search
Search

TAOK1| Gene Symbol: TAOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301247.2 | NP_001288176.1 | 6 - 7 | 726 | 89 | |
| XM_023651621.1 | XP_023507389.1 | 850 | 89 | |||
| XM_023651622.1 | XP_023507390.1 | 908 | 89 | |||
| GenBank mRNA | JL626564.1 | - | 4 - 5 | 362 | 89 | |
Set Membership: |
|