Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ec04950782_m1
See other LRRFIP2 GE Assays ›
Gene Symbol:
LRRFIP2
Gene Name:
LRR binding FLII interacting protein 2
Gene Aliases:
-
Chromosome Location:
Chr.16: 49545352 - 49654021 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID
Ec04950782_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: LRRFIP2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001308938.1
NP_001295867.1
11 - 12
1144
65
XM_001489140.6
XP_001489190.4
2441
65
XM_003363140.4
XP_003363188.1
1412
65
XM_003363141.4
XP_003363189.1
1484
65
XM_005600823.3
XP_005600880.1
2264
65
XM_005600824.3
XP_005600881.1
2308
65
XM_005600826.3
XP_005600883.1
2369
65
XM_005600827.3
XP_005600884.1
2348
65
XM_005600828.3
XP_005600885.1
2333
65
XM_005600829.3
XP_005600886.1
2246
65
XM_014731469.2
XP_014586955.1
2399
65
XM_014731470.2
XP_014586956.1
2390
65
XM_014731471.2
XP_014586957.1
2177
65
XM_014731472.2
XP_014586958.1
2174
65
XM_014731473.2
XP_014586959.1
2009
65
XM_023619767.1
XP_023475535.1
2309
65
XM_023619768.1
XP_023475536.1
2198
65
XM_023619769.1
XP_023475537.1
2372
65
XM_023619770.1
XP_023475538.1
2366
65
XM_023619771.1
XP_023475539.1
2351
65
XM_023619772.1
XP_023475540.1
2330
65
XM_023619773.1
XP_023475541.1
2324
65
XM_023619774.1
XP_023475542.1
2291
65
XM_023619775.1
XP_023475543.1
2288
65
XM_023619776.1
XP_023475544.1
2279
65
XM_023619777.1
XP_023475545.1
2276
65
XM_023619778.1
XP_023475546.1
2273
65
XM_023619779.1
XP_023475547.1
2258
65
XM_023619780.1
XP_023475548.1
2246
65
XM_023619782.1
XP_023475550.1
2231
65
XM_023619783.1
XP_023475551.1
2225
65
XM_023619784.1
XP_023475552.1
2222
65
XM_023619785.1
XP_023475553.1
2057
65
XM_023619786.1
XP_023475554.1
2009
65
XM_023619787.1
XP_023475555.1
1823
65
XM_023619788.1
XP_023475556.1
1688
65
XM_023619789.1
XP_023475557.1
1679
65
XM_023619790.1
XP_023475558.1
1546
65
XM_023619791.1
XP_023475559.1
1616
65
XM_023619792.1
XP_023475560.1
1607
65
XM_023619793.1
XP_023475561.1
1586
65
XM_023619794.1
XP_023475562.1
1514
65
XM_023619795.1
XP_023475563.1
2114
65
XM_023619796.1
XP_023475564.1
2207
65
GenBank mRNA
JL616096.1
-
11 - 12
1145
65
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Recommended Assay
Equus caballus
Probe spans exons
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline