Search
Search

CELF1| Gene Symbol: CELF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309238.1 | NP_001296167.1 | 11 - 12 | 1202 | 62 | |
| XM_023653793.1 | XP_023509561.1 | 1280 | 62 | |||
| XM_023653794.1 | XP_023509562.1 | 1280 | 62 | |||
| XM_023653795.1 | XP_023509563.1 | 1280 | 62 | |||
| XM_023653796.1 | XP_023509564.1 | 1277 | 62 | |||
| XM_023653797.1 | XP_023509565.1 | 1440 | 62 | |||
| XM_023653798.1 | XP_023509566.1 | 1166 | 62 | |||
| XM_023653799.1 | XP_023509567.1 | 1157 | 62 | |||
| XM_023653800.1 | XP_023509568.1 | 1298 | 62 | |||
| XM_023653802.1 | XP_023509570.1 | 1298 | 62 | |||
| XM_023653803.1 | XP_023509571.1 | 1154 | 62 | |||
| XM_023653804.1 | XP_023509572.1 | 1287 | 62 | |||
| XM_023653805.1 | XP_023509573.1 | 1362 | 62 | |||
| XM_023653806.1 | XP_023509574.1 | 1252 | 62 | |||
Set Membership: |
|