Search
Search
| Gene Symbol: GBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309252.1 | NP_001296181.1 | 6 - 7 | 1165 | 92 | |
| XM_005610442.3 | XP_005610499.2 | 1705 | 92 | |||
| XM_014739997.2 | XP_014595483.2 | 1414 | 92 | |||
| XM_014739998.2 | XP_014595484.2 | 1743 | 92 | |||
Set Membership: |
|